Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody (29E4.G7), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB001965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001965 Supplier Novus Biologicals Supplier No. NBP197410AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 29E4.G7
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Mouse
Immunogen This DYKDDDDK Epitope Tag Antibody (29E4.G7) was produced in mice by repeated immunizations with a synthetic peptide corresponding to the FLAG(TM) epitope tag peptide DYKDDDDK Epitope Tag Epitope Tag (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) conjugated to KLH using maleimide. Residues of glycine and cysteine were added to the termini to facilitate coupling.
Molecular Weight of Antigen 1.01 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity The purified antibody is directed against the FLAG(TM) motif and is useful in determining its presence in various assays where the epitope tag is present at either the amino or carboxy terminus of recombinant proteins. This monoclonal anti-FLAG(TM) tag antibody detects over-expressed proteins containing the FLAG(TM) epitope tag. In western blotting of bacterial extracts, the antibody does not cross-react with endogenous proteins.
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.