Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody (2F11G1), Novus Biologicals™
SDP

Catalog No. NBP237680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237680 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237680 Supplier Novus Biologicals Supplier No. NBP237680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F11G1
Conjugate Unconjugated
Dilution Western Blot 1:500- 1:2000, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Mouse
Immunogen DYKDDDDK Epitope Tag Antibody (2F11G1) was made to a Synthetic peptide corresponding to aa(DYKDDDDK), conjugated to KLH.
Molecular Weight of Antigen 1.01 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.