Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody (F-tag-01), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB500554AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500554AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500554AI Supplier Novus Biologicals Supplier No. NB500554AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone F-tag-01
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Mouse
Immunogen DYKDDDDK Epitope Tag Antibody (F-tag-01) was made to a Synthetic peptide: DYKDDDDK conjugated to KLH.
Molecular Weight of Antigen 1.01 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity DYKDDDDK Epitope Tag Antibody (F-tag-01) recognizes DDDDK-tagged proteins in all species. The small size of this tag and its high hydrophilicity decrease the probability of interference with its expression, proteolytic maturation, antigenicity, localization and function.
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.