Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody (F-tag-01), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359142 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359142 Supplier Novus Biologicals Supplier No. NB500554MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F-tag-01
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Mouse
Immunogen DYKDDDDK Epitope Tag Antibody (F-tag-01) was made to a Synthetic peptide: DYKDDDDK conjugated to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.