Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Epitope Tag Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB600344F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600344F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600344F Supplier Novus Biologicals Supplier No. NB600344F
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DYKDDDDK Epitope Tag Polyclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen DYKDDDDK Epitope Tag
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Goat
Immunogen To produce DYKDDDDK Epitope Tag Antibody, Goats were immunized with ECS, Enterokinase Cleavage Site (xxxDDDDK), conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Molecular Weight of Antigen 1.01 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity DYKDDDDK Epitope Tag Antibody reacts with FLAG.
Content And Storage Store at 4C in the dark.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.