Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23783350UG
Description
DYKDDDDK Epitope Tag Polyclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunofluorescence.Specifications
DYKDDDDK Epitope Tag | |
Polyclonal | |
Unconjugated | |
PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
Rabbit | |
1.01 kDa | |
50 μg | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. | |
Epitope Tag |
Western Blot, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:500, Immunofluorescence 1 μg/mL | |
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
DYKDDDDK Epitope Tag Antibody was made to a Synthetic Peptide: D Y K D D D D K C | |
Affinity Purified | |
RUO | |
DYKDDDDK Epitope Tag Antibody detects the DYKDDDDK epitope tag sequence. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction