Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | DYKDDDDK Epitope Tag |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DYKDDDDK Epitope Tag Polyclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunofluorescence.Specifications
DYKDDDDK Epitope Tag | |
Western Blot, Immunofluorescence | |
Unconjugated | |
RUO | |
DYKDDDDK Epitope Tag Antibody was made to a Synthetic Peptide: D Y K D D D D K C | |
Primary | |
DYKDDDDK Epitope Tag Antibody detects the DYKDDDDK epitope tag sequence. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
IgG | |
Affinity Purified | |
1.01 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title