Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynactin Subunit 2/DCTN2/DCTN-50 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB354180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354180 Supplier Novus Biologicals Supplier No. NBP297257MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dynactin Subunit 2/DCTN2/DCTN-50 Polyclonal antibody specifically detects Dynactin Subunit 2/DCTN2/DCTN-50 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Dynactin Subunit 2/DCTN2/DCTN-50
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DCTN-50, DCTN5050 kD dynein-associated polypeptide, dynactin 2 (p50), dynactin complex 50 kD subunit, Dynactin complex 50 kDa subunit, dynactin subunit 2,50 kDa dynein-associated polypeptide, DYNAMITIN, p50 dynamitin, RBP50
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Dynactin Subunit 2/DCTN2/DCTN-50 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10540
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.