Dynalon™ Bio-Bin™ Waste Disposal Containers

A new cost effective and convenient alternative to rigid plastic waste collection and disposal containers

Manufacturer: Dynalon™ 7973030006

 View more versions of this product

Catalog No. 12-009-801

  Check Availability
Add to Cart

    Specifications

    Capacity 1.5 gal., 6L
    Dimensions (L x W x H) 6 x 6 x 20.75 in. (15.24 x 15.24 x 52.67cm)
    For Use With (Application) Ideal solution for biohazard or non-hazardous waste
    Includes 2 x 2 in. biohazard waste labels
    Length (Metric) 15.24cm
    Width (Metric) 15.24cm
    Capacity (English) 1.5 gal.
    Capacity (Metric) 6L
    Height (English) 20.75 in.
    Height (Metric) 52.67cm
    Product Type Waste Container
    Certifications/Compliance UN 3291
    Length (English) 6 in.
    Width (English) 6 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit