Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin Antibody (D5), Novus Biologicals™
SDP

Catalog No. NB1001993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001993 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001993 Supplier Novus Biologicals Supplier No. NB1001993
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dynamin Monoclonal specifically detects Dynamin in Bovine, Chicken, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Dynamin
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 mg/mL, Flow Cytometry 10 ug/ml, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50
Gene Accession No. Q05193
Gene Alias DNM, dynamin 1, dynamin-1, EC 3.6.5.5
Gene Symbols DNM1
Host Species Mouse
Immunogen Recombinant His6-tagged human dynamin.
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1759
Test Specificity Cross-reacts with Human, Mouse, Rat, Cow and Chicken. Not yet tested in other species.
Target Species Bovine, Chicken, Rat, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.