Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin Antibody, Novus Biologicals™
SDP

Catalog No. NB300565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300565 Supplier Novus Biologicals Supplier No. NB300565

Rabbit Polyclonal Antibody

Dynamin Polyclonal specifically detects Dynamin in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Dynamin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry 1:10 - 1:500
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P21575
Gene Alias DNM, dynamin 1, dynamin-1, EC 3.6.5.5
Gene Symbols DNM1
Host Species Rabbit
Immunogen Synthetic Peptide corresponding to resdiues C K(633) E K A S E T E E N G S D S F(647) of rat Dyn1.
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1759
Test Specificity Dynamin 1 - Synapse
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.