Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin, p Ser774 Antibody, Novus Biologicals™
SDP

Catalog No. NB300209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300209 Supplier Novus Biologicals Supplier No. NB300209
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Dynamin Polyclonal specifically detects Dynamin in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Dynamin (p Ser774)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P21575
Gene Alias DNM, dynamin 1, dynamin-1, EC 3.6.5.5
Gene Symbols DNM1
Host Species Sheep
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser774 conjugated to KLH. Accession # P21575
Molecular Weight of Antigen 95 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1759
Test Specificity Specific for the ∼95k dynamin protein phosphorylated at Ser774. Labels the purified protein phosphorylated in vitro by cdk5 but not by PKC. Does not cross react with other purified substrates of cdk5 (e.g. amphiphysin and synapsin). Immunolabeling is blocked by lambda-phosphatase treatment.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.