Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Dynapyrazole A

Click to view available options
Quantity:
5 mg
25 mg
Description
Dynapyrazole A is a dynein inhibitor. Inhibits dynein 1 and dynein 2-dependent microtubule gliding (IC50 values are 2.3 and 2.6 μM, respectively). Inhibits hedgehog signaling in vitro (IC50 = 1.9 μM). Displays reversible inhibition of retrograde and anterograde intraflagellar transport in Chlamydomonas; displays stronger inhibition of retrograde transport.
Specifications
Specifications
| Chemical Name or Material | 2-[1-(4-Chlorophenyl)cyclopropyl]-4,5-dihydro-7-iodo-5-oxopyrazolo[1,5-a]quinazoline-3-carbonitrile |
| CAS | 2226517-75-3 |
| Quantity | 25 mg |
| Target | Hedgehog Signaling Inhibitors |
| Molecular Formula | C20H12ClIN4O |
| Formula Weight | 3333 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction