Learn More
Dynatech Precision Sampling Heavy Duty Purge and Trap Syringe from Restek
SDP
Catalog No. 06711463 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Includes:
Not Applicable
Sample lok valve
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Includes
06-711-463 5 mL Sample lok valve
06-711-462 10 mL Sample lok valve
06-711-464 10 mL Not Applicable
06-711-461 5 mL Not Applicable
06-711-460 2 mL Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-711-463 Supplier Restek Corporation Supplier No. 21208
Add to Cart
Add to Cart
Heavy-duty glass barrel with metal front and rear flanges.
  • PTFE luer lock tip
  • Can fill and empty sparge tubes
  • Sample-Lok valve accepts standard luer lock needles
  • Suitable for purge and trap applications
Specifications
Material PTFE (Tip)
For Use With (Application) For purge and trap applications
Includes Sample lok valve
Product Type Syringe
Quantity Ea.
Volume (Metric) 5 mL
Connection Type Luer Lock