Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYNC1LI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167207 100 μg
NB167208 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167207 Supplier Novus Biologicals Supplier No. NBP316056100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYNC1LI2 Polyclonal antibody specifically detects DYNC1LI2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DYNC1LI2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 50% glycerol, pH7.3
Gene Alias cytoplasmic dynein 1 light intermediate chain 2, DNCLI2, Dynein light intermediate chain 2, cytosolic, dynein, cytoplasmic 1, light intermediate chain 2, dynein, cytoplasmic, light intermediate polypeptide 2, LIC2, LIC-2, LIC53/55
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 328-492 of human DYNC1LI2 (NP_006132.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1783
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.