Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynein intermediate chain 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Dynein intermediate chain 2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18050920
![]() |
Novus Biologicals
NBP18050920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180509
![]() |
Novus Biologicals
NBP180509 |
100 μL |
Each for $487.50
|
|
|||||
Description
Dynein intermediate chain 2 Polyclonal specifically detects Dynein intermediate chain 2 in Human samples. It is validated for Western Blot.Specifications
Dynein intermediate chain 2 | |
Polyclonal | |
Rabbit | |
NP_075462 | |
64446 | |
Synthetic peptide directed towards the N terminal of human DNAI2. Peptide sequence TRGVNHVEGGWPKDVNPLELEQTIRFRKKVEKDENYVNAIMQLGSIMEHC. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Axonemal dynein intermediate chain 2, CILD9, dynein intermediate chain 2, axonemal, dynein, axonemal, intermediate chain 2, dynein, axonemal, intermediate polypeptide 2 | |
DNAI2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title