Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYSFIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155526
Description
DYSFIP1 Polyclonal specifically detects DYSFIP1 in Human samples. It is validated for Western Blot.Specifications
| DYSFIP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dysferlin interacting protein 1, dysferlin interacting protein 1 (toonin), dysferlin-interacting protein 1, dysferlin-interacting protein 1 (toonin), MGC138299, toonin | |
| Rabbit | |
| 17 kDa | |
| 100 μL | |
| Primary | |
| Guinea pig 86%, Equine 86%, Porcine 86%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q86WC6 | |
| PPP1R27 | |
| Synthetic peptides corresponding to DYSFIP1(dysferlin interacting protein 1) The peptide sequence was selected from the middle region of DYSFIP1. Peptide sequence CSDGYPDIARYLISLGADRDATNDDGDLPSDLIDPDYKELVELFKGTTMD. | |
| Affinity purified | |
| RUO | |
| 116729 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction