Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYSFIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | DYSFIP1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DYSFIP1 Polyclonal specifically detects DYSFIP1 in Human samples. It is validated for Western Blot.Specifications
| DYSFIP1 | |
| Polyclonal | |
| Rabbit | |
| Q86WC6 | |
| 116729 | |
| Synthetic peptides corresponding to DYSFIP1(dysferlin interacting protein 1) The peptide sequence was selected from the middle region of DYSFIP1. Peptide sequence CSDGYPDIARYLISLGADRDATNDDGDLPSDLIDPDYKELVELFKGTTMD. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| dysferlin interacting protein 1, dysferlin interacting protein 1 (toonin), dysferlin-interacting protein 1, dysferlin-interacting protein 1 (toonin), MGC138299, toonin | |
| PPP1R27 | |
| IgG | |
| 17 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title