Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Catalog No. AA1459522
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA1459522

Supplier: Thermo Scientific Chemicals 01459522

Only null left
Add to Cart

CAS: 15280-55-4 | C6H17DyO10 | 411.69 g/mol

Dysprosium Aceate has specialized uses in ceramics, glass, phosphors, lasers and Dysprosium Metal halide lamp. High purity of Dysprosium Aceate is used in electronics industry as an antireflection coating in photoelectric devices. Dysprosium is used in conjunction with vanadium and other elements, in making laser materials and commercial lighting. Dysprosium and its compounds are highly susceptible to magnetization, they are employed in various data-storage applications, such as in hard disks. It is also used in dosimeters for measuring ionizing radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-55-4
Molecular Formula C6H17DyO10
Molecular Weight (g/mol) 411.69
MDL Number MFCD00013033
InChI Key SIYZPSYZNCQFDV-UHFFFAOYSA-K
IUPAC Name dysprosium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Dy+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline Aggregates
Quantity 100 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 411.68 (339.62 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Assay (REO)
Chemical Name or Material Dysprosium(III) acetate tetrahydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.