Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystonin Antibody (H5E.Hy.4B), Novus Biologicals™
SDP

Catalog No. DDX0450P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0450P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0450P100 Supplier Novus Biologicals Supplier No. DDX0450P100
Add to Cart
Add to Cart

Human Monoclonal Antibody

Dystonin Monoclonal specifically detects Dystonin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Dystonin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone H5E.Hy.4B
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias 230 kDa bullous pemphigoid antigen, BP240, BPAFLJ30627, BPAG1FLJ32235, Bullous pemphigoid antigen, Bullous pemphigoid antigen 1, bullous pemphigoid antigen 1, 230/240kDa, CATX-15, D6S1101, DKFZp564B2416, DMHCATX15, DT, Dystonia musculorum protein, dystonin, FLJ13425, FLJ21489, FLJ46791, Hemidesmosomal plaque protein, KIAA0465, KIAA0728, KIAA1470, MACF2, trabeculin-beta
Gene Symbols DST
Host Species Human
Immunogen Bullous Pemphigoid Antigen was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 667
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.