Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrobrevin alpha Antibody (OTI1B2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP270591IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270591IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270591IR Supplier Novus Biologicals Supplier No. NBP270591IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dystrobrevin alpha
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B2
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1
Gene Symbols DTNA
Host Species Mouse
Immunogen Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1837.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.