Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrophin Antibody (DMD/3677), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB379030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379030 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379030 Supplier Novus Biologicals Supplier No. NBP308707UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Dystrophin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DMD/3677
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
Host Species Mouse
Immunogen A recombinant fragment (around aa 114-263) of human DMD protein (exact sequence is proprietary) (Uniprot: P11532)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1756
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.