Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DZIP1 Monoclonal Antibody (05), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | DZIP1 |
---|---|
Clone | 05 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543523
|
Invitrogen™
MA543523 |
50 μL |
Each for $285.50
|
|
|||||
PIMA542965
|
Invitrogen™
MA542965 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human DZIP1
May participate in spermatogenesis via its interaction with DAZ.Specifications
DZIP1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
2510025K24Rik; 2810422M04Rik; DAZ interacting protein 1; DAZ interacting protein testis1; DAZ interacting zinc finger protein 1; DAZ-interacting protein 1 homolog; DAZ-interacting protein 1/2; DZIP; Dzip1; DZIP2; DZIPt1; KIAA0996; zinc finger DAZ interacting protein 1; zinc finger protein DZIP1; zinc-finger protein DZIPt1 | |
Dzip1 | |
IgG2a | |
Protein A | |
Antibody |
05 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q86YF9 | |
22873 | |
A synthetic peptide corresponding to the center region of the Human DZIP1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Dzip1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title