Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DZIP3 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB044807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044807 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044807 Supplier Novus Biologicals Supplier No. NBP297549UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DZIP3 Polyclonal specifically detects DZIP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DZIP3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DAZ interacting protein 3, zinc finger, DAZ-interacting protein 3, E3 ubiquitin-protein ligase DZIP3, EC 6.3.2.-, FLJ13327, FLJ57977, FLJ58022, FLJ58223, hRUL138KIAA0675, human RNA-binding ubiquitin ligase of 138 kDa, RNA-binding ubiquitin ligase of 138 kDa, UURF2, UURF2 ubiquitin ligase, zinc finger DAZ interacting protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DZIP3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9666
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.