Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ E-4031 dihydrochloride
GREENER_CHOICESDP

Catalog No. 180810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-081-0 10 mg
18-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-081-0 Supplier Tocris Bioscience™ Supplier No. 1808/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV11.1 (hERG) channel blocker; inhibits rapid delayed rectifier K+ current (IKr)

Selective blocker of KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Reversibly prolongs action potential duration in guinea pig papillary muscle and isolated ventricular myocytes, without affecting Na+ or Ca2+ inward currents. Class III antiarrhythmic agent.

Chemical Identifiers

CAS 113559-13-0
Molecular Formula C21H29Cl2N3O3S

Specifications

Quantity 10 mg
Formula Weight 474.44
Purity >98%
Molecular Weight (g/mol) 474.44
Chemical Name or Material E-4031 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.