Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Antibody (67A4), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP142793J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142793J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142793J Supplier Novus Biologicals Supplier No. NBP142793APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen E-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone 67A4
Conjugate APC
Dilution Flow Cytometry
Gene Accession No. P12830
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Gene Symbols CDH1
Host Species Mouse
Immunogen T-47D cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 999
Test Specificity The mouse monoclonal antibody 67A4 recognizes CD324 E-cadherin, an approximately 100 kDa epithelial cell adhesion molecule, whose detection is important for determination of invasive potential of epithelial neoplasms. HLDA VIII
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.