Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Antibody (967717), Alexa Fluor 350, Novus Biologicals™
SDP

Catalog No. MB18384A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB18384A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB18384A350 Supplier Novus Biologicals Supplier No. MAB18384AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Immunocytochemistry, Western Blot.

Specifications

Antigen E-Cadherin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 967717
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Gene Symbols CDH1
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human E-Cadherin
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 999.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.