Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Cadherin Antibody (967738), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314063 Supplier Novus Biologicals Supplier No. FAB18383PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen E-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone 967738
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human E-Cadherin, Asp155-Ile707, Accession # P12830
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 999
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.