Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli 015 Antibody (15/443) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064541 0.1 mg
NB017599 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064541 Supplier Novus Biologicals Supplier No. NB10064541
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E. Coli 015 Monoclonal specifically detects E. Coli 015 in Bacteria samples. It is validated for ELISA.

Specifications

Antigen E. Coli 015
Applications ELISA
Classification Monoclonal
Clone 15/443
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias Escherichia coli
Host Species Mouse
Immunogen Native, NCTC 9015
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In a simple ELISA using a number of Biogenesis Test Groups, this is only reactive with E. coli 015 (NCTC 9015). Reactive with isolates fixed by phenol, formalin and boiling.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.