Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli 015 Antibody (15/443) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$170.97 - $492.89
Specifications
| Antigen | E. Coli 015 |
|---|---|
| Clone | 15/443 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Immunocytochemistry, Immunofluorescence, ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017599
![]() |
Novus Biologicals
NB100645410.025MG |
0.025 mg |
Each for $170.97
|
|
|||||
NB10064541
![]() |
Novus Biologicals
NB10064541 |
0.1 mg |
Each for $492.89
|
|
|||||
Description
E. Coli 015 Monoclonal specifically detects E. Coli 015 in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| E. Coli 015 | |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Native, NCTC 9015 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 15/443 | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Escherichia coli | |
| IgG3 | |
| Protein A purified | |
| In a simple ELISA using a number of Biogenesis Test Groups, this is only reactive with E. coli 015 (NCTC 9015). Reactive with isolates fixed by phenol, formalin and boiling. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title