Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli 987P Pili Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062530 0.5 mL
NB017490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062530 Supplier Novus Biologicals Supplier No. NB10062530

Sheep Polyclonal Antibody

E. Coli 987P Pili Polyclonal specifically detects E. Coli 987P Pili in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E. Coli 987P Pili
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 9.7 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P21413
Gene Alias Escherichia coli
Host Species Sheep
Immunogen E. coli strain CVL Moon 987P O:9K103 was used.
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes the E. coli 987P pilus, a structure that mediates attachment and subsequent colonization of host surfaces.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.