Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli 987P Pili Antibody - BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | E. Coli 987P Pili |
---|---|
Concentration | 9.7 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
Applications | Immunofluorescence, ELISA, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017490
![]() |
Novus Biologicals
NB100625300.1ML |
0.1 mL |
Each for $206.00
|
|
|||||
NB10062530
![]() |
Novus Biologicals
NB10062530 |
0.5 mL |
Each for $487.50
|
|
|||||
Description
E. Coli 987P Pili Polyclonal specifically detects E. Coli 987P Pili in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
E. Coli 987P Pili | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Purified | |
RUO | |
Phosphate buffered saline with 0.09% Sodium Azide | |
Escherichia coli | |
IgG | |
IgG purified | |
This product recognizes the E. coli 987P pilus, a structure that mediates attachment and subsequent colonization of host surfaces. |
9.7 mg/mL | |
Immunofluorescence, ELISA, Immunocytochemistry | |
Unconjugated | |
Sheep | |
Bacteria | |
P21413 | |
E. coli strain CVL Moon 987P O:9K103 was used. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title