Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB001703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001703 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001703 Supplier Novus Biologicals Supplier No. NB10062528500UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E. Coli Polyclonal specifically detects E. Coli in Bacteria samples. It is validated for ELISA.

Specifications

Antigen E. Coli
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Dilution ELISA 1:100-1:2000
Formulation PBS
Gene Alias Escherichia coli
Host Species Rabbit
Immunogen A mixture of all antigenic serotypes.
Quantity 500 μL
Test Specificity NB100-62526 reacts with all 'O' and 'K' antigenic serotypes of E. coli. This product has been used versus serotypes O157:H7, O20, O125, 055, 0111 and K12 and will remove E. coli proteins from recombinant preparations. This antibody is not absorbed and cross-reacts with related Enterobacteriaceae
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.