Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
$816.50
Specifications
| Antigen | E. Coli |
|---|---|
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
Description
E. Coli Polyclonal specifically detects E. Coli in Bacteria samples. It is validated for ELISA.Specifications
| E. Coli | |
| ELISA | |
| Biotin | |
| Rabbit | |
| PBS | |
| A mixture of all antigenic serotypes. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| Bacteria | |
| Escherichia coli | |
| IgG | |
| NB100-62526 reacts with all 'O' and 'K' antigenic serotypes of E. coli. This product has been used versus serotypes O157:H7, O20, O125, 055, 0111 and K12 and will remove E. coli proteins from recombinant preparations. This antibody is not absorbed and cross-reacts with related Enterobacteriaceae |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title