Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100644470.1ML
Description
E. Coli Polyclonal specifically detects E. Coli in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
E. Coli | |
Polyclonal | |
Unconjugated | |
Phosphate buffered saline with 0.1% Sodium Azide | |
Goat | |
IgG purified | |
RUO | |
Recognises many “O” and “K” antigenic serotypes of Escherichia coli. The antiserum is unabsorbed and cross reacts with related Enterobacteriaceae. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence, Immunocytochemistry | |
4.0 mg/mL | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Escherichia coli | |
A mixture of E.coli serotypes | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction