Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. Coli Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | E. Coli |
---|---|
Concentration | 4.0 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017581
![]() |
Novus Biologicals
NB100644470.1ML |
0.1 mL |
Each for $181.90
|
|
|||||
NB10064447
![]() |
Novus Biologicals
NB10064447 |
0.5 mL |
Each for $487.50
|
|
|||||
Description
E. Coli Polyclonal specifically detects E. Coli in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
E. Coli | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate buffered saline with 0.1% Sodium Azide | |
A mixture of E.coli serotypes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4.0 mg/mL | |
Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Goat | |
Bacteria | |
Escherichia coli | |
IgG | |
IgG purified | |
Recognises many “O” and “K” antigenic serotypes of Escherichia coli. The antiserum is unabsorbed and cross reacts with related Enterobacteriaceae. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title