Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli J5 LPS Antibody (2D7/1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10064543S Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064543S 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064543S Supplier Novus Biologicals Supplier No. NB10064543R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E. Coli J5 LPS Monoclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen E. Coli J5 LPS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D7/1
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Escherichia coli
Host Species Mouse
Immunogen E. coli J5 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This is specific for E. coli J5 LPS. It has also been found to react with K. pneumoniae, S. sonnei and S. typhimurium.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.