Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli J5 LPS Antibody, Novus Biologicals™
SDP

Catalog No. NB10062551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062551 0.5 mL
NB017495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062551 Supplier Novus Biologicals Supplier No. NB10062551
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

E. Coli J5 LPS Polyclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for ELISA.

Specifications

Antigen E. Coli J5 LPS
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation Whole antisera with 0.09% Sodium Azide
Gene Alias Escherichia coli
Host Species Sheep
Immunogen Heat inactivated Escherichia coli J5 (Rc mutant; ATCC)
Purification Method Unpurified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62551 recognizes the J5 mutant of E. coli 0:111 which lacks the enzyme uridine diphosphate glucose 4-epimerase and therefore produces an incomplete LPS, deficient in galactose and all the sugars distal to the central polymers. The 55 mutant has no O-specific chains and its endotoxin remains as the core LPS containing lipid A, N acetyl glucosamine, 2-keto-3-deoxyoctonate, heptose and glucose (a composition similar to that of the Rc strains of Salmonella).
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.