Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli K88A Antibody (1050/66), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10064537Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064537Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064537Y Supplier Novus Biologicals Supplier No. NB10064537AF647

Mouse Monoclonal Antibody

E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E. Coli K88A
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1050/66
Conjugate Alexa Fluor 647
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Escherichia coli
Gene Symbols faeC
Host Species Mouse
Immunogen NCTC 10964 (Nobivac)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling.
Reconstitution Protein A purified
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.