Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli K88A Antibody (1050/66), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB064537549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064537549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064537549 Supplier Novus Biologicals Supplier No. NB10064537JF549

Mouse Monoclonal Antibody

E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E.ColiK88A
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1050/66
Conjugate Janelia Fluor 549
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Escherichia coli
Gene Symbols faeC
Host Species Mouse
Immunogen NCTC 10964 (Nobivac)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.