Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli K88AB Pili Antibody, Novus Biologicals™
SDP

Catalog No. NB10062532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062532 0.5 mL
NB017491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062532 Supplier Novus Biologicals Supplier No. NB10062532
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

E. Coli K88AB Pili Polyclonal specifically detects E. Coli K88AB Pili in Bacteria samples. It is validated for ELISA.

Specifications

Antigen E. Coli K88AB Pili
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.1% Sodium Azide
Gene Accession No. P02970
Gene Alias Escherichia coli
Gene Symbols faeG
Host Species Sheep
Immunogen Escherichia colii strain 08:K87:K88ab
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity ESCHERICHIA COLI K88AB PILI
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.