Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli Labile Toxin A Chain Antibody (20/480), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10064577Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064577Z 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064577Z Supplier Novus Biologicals Supplier No. NB10064577AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E. Coli Labile Toxin A Chain Monoclonal specifically detects E. Coli Labile Toxin A Chain in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen E. Coli Labile Toxin A Chain
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 20/480
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Escherichia coli
Gene Symbols eltA
Host Species Mouse
Immunogen Native, labile toxin
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli labile toxin, C terminal of A chain.
Reconstitution Protein A purified
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.