Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli O157 Antibody (9.88), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB008071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008071 Supplier Novus Biologicals Supplier No. NBP254353AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E. Coli O157 Monoclonal specifically detects E. Coli O157 in E. coli samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E. Coli O157
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9.88
Conjugate Alexa Fluor 594
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen A crude sonicate of E. coli O157
Purification Method Protein L or PEG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody shows specificity to E.coli O157 in a simple ELISA. Escherichia coli are Gram negative bacterium that are commonly found in the lower intestine of warm-blooded organisms (endotherms). Their serological types are determined in combination with somatic antigens (O group: O1-O173) and flagella antigens (H type: H1-H56). TheE. coli that cause intestinal infectious diseases including diarrhea, acute gastritis or colitis are referred to as pathogenicE. coli, which are classified into the following 4 groups according to differences in the mode of pathogenicity; enteropathogenicE. coli(EPEC), enteroinvasiveE. coli(EIEC), enterotoxigenicE. coli(ETEC) and enterohemorrhagicE. coli(EHEC). Although the identification of pathogenicE. coli requires verification of their pathogenicity, pathogenicE. coli often have specific serotypes; therefore, typing of the serogroup and serotype is necessary in screening pathogenicE. coli.
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.