Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli O157 Antibody (9.88) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254353200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254353200 200 μg
NB254353100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254353200 Supplier Novus Biologicals Supplier No. NBP254353200UG

Mouse Monoclonal Antibody

E. Coli O157 Monoclonal antibody specifically detects E. Coli O157 in E. coli samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E. Coli O157
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9.88
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Formulation PBS with No Preservative
Host Species Mouse
Immunogen A crude sonicate of E. coli O157
Purification Method Tissue culture supernatant
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.