Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. Coli O157 Mouse anti-E. coli, Biotin, Clone: 9.88, Novus Biologicals™
SDP

Catalog No. NB019724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019724 Supplier Novus Biologicals Supplier No. NBP254353B0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E. Coli O157 Monoclonal specifically detects E. Coli O157 in E. coli samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen E. Coli O157
Applications Immunofluorescence
Classification Monoclonal
Clone 9.88
Conjugate Biotin
Formulation PBS
Host Species Mouse
Immunogen A crude sonicate of E. coli O157
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.