Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Antibody (105), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB315821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315821 Supplier Novus Biologicals Supplier No. NBP290951JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Cynomolgus Monkey samples. It is validated for ELISA

Specifications

Antigen E-Selectin/CD62E
Applications ELISA
Classification Monoclonal
Clone 105
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus E-Selectin/CD62E (Uniprot#: G8F370; Met1-Pro556).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6401
Target Species Cynomolgus Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.