Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Antibody (40), Alexa Fluor™ 532
SDP

Catalog No. NB107584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB107584 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB107584 Supplier Novus Biologicals Supplier No. NBP289430AF532

Rabbit Monoclonal Antibody

E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.

Specifications

Antigen E-Selectin/CD62E
Applications Western Blot, ELISA
Classification Monoclonal
Clone 40
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human E-Selectin/CD62E (NP_000441.2; Met 1-Pro 556).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity No cross-reactivity in ELISA with: Human CD31/ PECAM1 Human VCAM1/ CD106 Human ICAM1/ CD54 Mouse VCAM1/ CD106 Mouse ICAM-1/ CD54
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.