Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin/CD62E Antibody (P2H3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N143393PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N143393PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N143393PEC7 Supplier Novus Biologicals Supplier No. NBP143393PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen E-Selectin/CD62E
Applications Flow Cytometry
Classification Monoclonal
Clone P2H3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Gene Symbols SELE
Host Species Mouse
Immunogen The immunogen for this antibody was CD62E.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6401.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.