Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ E-Selectin/CD62E Antibody Pair [HRP]
SDP

Catalog No. p-7183122 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat E-Selectin/CD62E.

Specifications

Assay Range 18.8 pg/mL
Assay Sensitivity 18.8-1200 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat E-Selectin/CD62E.
Target Species Rat
For Use With (Application) Sandwich ELISA
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E
Gene ID (Entrez) 6401
Gene Symbol SELE
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL of rabbit anti-E-Selectin/CD62E monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 0.4 mg/mL of rabbit anti-Rat E-Selectin/CD62E monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 33 ng of recombinant Rat E-Selectin/CD62E.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Immunology, Lipid and Metabolism
    Storage Requirements Storage is content dependent.
    Target E-Selectin/CD62E
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1200 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.