Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Tag Epitope Tag Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB600525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600525 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600525 Supplier Novus Biologicals Supplier No. NB600525
Add to Cart
Add to Cart

Goat Polyclonal Antibody

E-Tag Epitope Tag Polyclonal antibody specifically detects E-Tag Epitope Tag in Epitope Tag samples. It is validated for Immunocytochemistry, Immunofluorescence, In vitro Assay, Immunohistochemistry.

Specifications

Antigen E-Tag Epitope Tag
Applications Immunocytochemistry, Immunofluorescence, In vitro Assay, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:400, In vitro assay, Immunohistochemistry (Negative)
Formulation PBS and 0.2% BSA with 0.09% Sodium Azide
Gene Alias E epitope tag, GAPVPYPDPLEPR epitope tag, GAPVPYPDPLEPR tag
Host Species Goat
Immunogen Goats were immunized with E-Tag (GAPVPYPDPLEPR) conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.